missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR2, Rat anti-Mouse, Clone: CL10, Novus Biologicals™
Rat Monoclonal Antibody
€ 288.00 - € 612.00
Specifications
| Antigen | FOLR2 |
|---|---|
| Clone | CL10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18032346
|
Novus Biologicals
NBP2-81079-0.025MG |
0.025 mg |
€ 288.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18084534
|
Novus Biologicals
NBP2-81079-0.2MG |
0.2 mg |
€ 612.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOLR2 Monoclonal specifically detects FOLR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| FOLR2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
| FOLR2 | |
| IgG2a κ | |
| Protein A purified |
| CL10 | |
| Western Blot, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 2350 | |
| FRbeta -transfected RBL2H3 cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title