Learn More
Poulten Graf™ FORTUNA™ Class B Low-Form Glass Measuring Cylinders
Brand:  Poulten Graf™ 1 3204502
Additional Details : Weight : 0.21000kg
 View more versions of this product
Product Code. 15942150
  • € 17.80 / Pack of 2
Estimated Shipment: 28-12-2022
to see stock.
Add to basket
Specifications
Graduated Measuring Cylinder
EN ISO 4788
Blue
Hexagonal
50 mL
2 mL
B
Round
To Contain
Borosilicate Glass
Blue Graduations
Autoclavable
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers