Learn More
FOXL2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 44 publications
Brand: Novus Biologicals NB100-1277
This item is not returnable.
View return policy
Description
FOXL2 Polyclonal specifically detects FOXL2 in Human, Mouse, Bovine, Ferret samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Whole-Mount.