Learn More
FOXL2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 44 publications
Brand:  Novus Biologicals NB100-1277
 View more versions of this product
Product Code. 18725683
  • € 631.00 / 0.10mg
Description
FOXL2 Polyclonal specifically detects FOXL2 in Human, Mouse, Bovine, Ferret samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Whole-Mount.