Learn More
FOXL2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 44 publications
€ 288.00 - € 631.00
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18725683
|
Novus Biologicals
NB100-1277 |
€ 631.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18590493
|
Novus Biologicals
NB100-1277-20ug |
20 μg |
€ 288.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXL2 Polyclonal specifically detects FOXL2 in Human, Mouse, Bovine, Ferret samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Whole-Mount.For Research Use Only