Learn More
FOXO4 Recombinant Rabbit Monoclonal Antibody (23GB1470), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen MA553994
 View more versions of this product
Product Code. 30087654
  • € 562.00 / 100µL
Description
FOXO4 Recombinant Monoclonal antibody specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FOXO4
Recombinant Monoclonal
Unconjugated
FOXO4
AFX; Afx1; Afxh; AFXzeta; Fkhr3; Fork head domain transcription factor AFX1; forkhead box O4; forkhead box protein O4; forkhead protein; FOXO4; FOXO4b; MGC120490; MLLT7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; OTTHUMP00000023497; RGD1561201
Rabbit
Affinity Chromatography
RUO
302415, 4303, 54601
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1470
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P98177, Q9WVH3
FOXO4
A synthesized peptide derived from human FoxO4 (450-505AA).