Learn More
FoxP2 Antibody (RAB-S249) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20155-0.2mg
 View more versions of this product
Product Code. 18694714
  • € 578.00 / 0.20mg
Description
FoxP2 Monoclonal antibody specifically detects FoxP2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FoxP2
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
93986
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
RAB-S249
Unconjugated
CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein
This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP2 protein under non-denaturing conditions.
0.2 mg