missing translation for 'onlineSavingsMsg'
Learn More

FoxP2 Antibody (RAB-S249) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

€ 244.00 - € 578.00

Specifications

Antigen FoxP2
Clone RAB-S249
Concentration 1 mg/mL
Applications Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18654904
View Documents
Novus Biologicals
NBP3-20155-0.025mg
0.025 mg
€ 244.00
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18694714
View Documents
Novus Biologicals
NBP3-20155-0.2mg
0.2 mg
€ 578.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FoxP2 Monoclonal antibody specifically detects FoxP2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications

Specifications

FoxP2
1 mg/mL
Monoclonal
Purified
RUO
PBS
93986
IgG κ
Protein A purified
RAB-S249
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Human
CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein
This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP2 protein under non-denaturing conditions.
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.