Learn More
Novus Biologicals™ FPRL2/FPR3 Antibody (7D9B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27173-0.025ml
 View more versions of this product
Product Code. 30030198
  • € 310.00 / 0.03mL
Description
FPRL2/FPR3 Monoclonal antibody specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FPRL2/FPR3
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Purified recombinant fragment of human FPRL2/FPR3 expressed in E. Coli.
0.025 mL
GPCR, Immunology
2359
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
7D9B4
Unconjugated
PBS
Mouse
Protein G purified