missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FPRL2/FPR3 Antibody (7D9B4), Novus Biologicals™
Mouse Monoclonal Antibody
€ 310.00 - € 534.00
Specifications
| Antigen | FPRL2/FPR3 |
|---|---|
| Clone | 7D9B4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030198
|
Novus Biologicals™
NBP3-27173-0.025ml |
0.025 mL |
€ 310.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029774
|
Novus Biologicals™
NBP3-27173-0.1ml |
0.1 mL |
€ 534.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FPRL2/FPR3 Monoclonal antibody specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| FPRL2/FPR3 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| GPCR, Immunology | |
| PBS | |
| 2359 | |
| IgG1 | |
| Protein G purified |
| 7D9B4 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I | |
| Purified recombinant fragment of human FPRL2/FPR3 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title