missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA2 Antibody (SR1769), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22255-100ul
This item is not returnable.
View return policy
Description
FRA2 Monoclonal antibody specifically detects FRA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| FRA2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| SR1769 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 | |
| FOS-like antigen 2, fos-related antigen 2, FRA-2, FRA2FLJ23306 | |
| A synthesized peptide derived from human FRA2 (Uniprot #: P15408) | |
| 100 μg | |
| Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| 2355 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction