missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA2 Antibody (SR1769), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | FRA2 |
|---|---|
| Clone | SR1769 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650784
|
Novus Biologicals
NBP3-22255-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619693
|
Novus Biologicals
NBP3-22255-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FRA2 Monoclonal antibody specifically detects FRA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
| FRA2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FOS-like antigen 2, fos-related antigen 2, FRA-2, FRA2FLJ23306 | |
| A synthesized peptide derived from human FRA2 (Uniprot #: P15408) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1769 | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2355 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title