Learn More
3M™ Fuel™ X2 Safety Glasses
Catalog Number 15739687
Click to view available options
Lens Tint:
Bronze
Clear
Red, Mirror
3M™ Fuel™ X2 Safety Glasses have a revolutionary frame design that features grooved styling, rubber over-molded adjustable temples, and a fully adjustable nose bridge.
  • Safety Glasses with a sleek half-frame profile helps ensure that protection and performance remain uncompromised
  • The frame design has bold new grooved styling and a fully adjustable nose bridge
  • Adjustable temple length for comfort and optimal fit
  • Optical class 1 for maximum clarity
  • Anti-Scratch and Anti-Fog coating for increased durability and improved vision
  • Frame made from a high density nylon material for impact, torsion and heat resistance
  • special mirror coating provides glare reduction and UV protection
  • Strong polycarbonate lens for impact resistance
Certifications/Compliance CE Approved, EN 166:2001
Description Red Mirror coating for increased durability and improved vision
Lens Tint Red, Mirror
Product Type Safety glasses
Frame Color Brown
Lens Material Polycarbonate