Learn More
3M™ Fuel™ X2 Safety Glasses
Click to view available options
Lens Tint:
Bronze
Clear
Red, Mirror
Description
- Safety Glasses with a sleek half-frame profile helps ensure that protection and performance remain uncompromised
- The frame design has bold new grooved styling and a fully adjustable nose bridge
- Adjustable temple length for comfort and optimal fit
- Optical class 1 for maximum clarity
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- Frame made from a high density nylon material for impact, torsion and heat resistance
- special mirror coating provides glare reduction and UV protection
- Strong polycarbonate lens for impact resistance
Specifications
Certifications/Compliance | CE Approved, EN 166:2001 |
Description | Red Mirror coating for increased durability and improved vision |
Lens Tint | Red, Mirror |
Product Type | Safety glasses |
Frame Color | Brown |
Lens Material | Polycarbonate |