Learn More
3M™ Fuel™ X2 Safety Glasses
Product Code. 15719687
Change view
Click to view available options
Lens Tint:
Bronze
Clear
Red, Mirror
Unit Size:
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint unitSize
15719687 Clear Pack of 20
15729687 Bronze Pack of 20
15739687 Red, Mirror Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15719687 Supplier 3M™ Supplier No. 7000032502
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Fuel™ X2 Safety Glasses have a revolutionary frame design that features grooved styling, rubber over-molded adjustable temples, and a fully adjustable nose bridge.
  • Safety Glasses with a sleek half-frame profile helps ensure that protection and performance remain uncompromised
  • The frame design has bold new grooved styling and a fully adjustable nose bridge
  • Adjustable temple length for comfort and optimal fit
  • Optical class 1 for maximum clarity
  • Anti-Scratch and Anti-Fog coating for increased durability and improved vision
  • Frame made from a high density nylon material for impact, torsion and heat resistance
  • special mirror coating provides glare reduction and UV protection
  • Strong polycarbonate lens for impact resistance
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, EN 166:2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety glasses
Frame Color Brown
Lens Material Polycarbonate