Learn More
Ekastu Safety™ CLARO Full-View Goggles
Description
- DIN EN 166 1 3 4 9 – BT
- Full view goggles made of soft plastic against dust and liquid splashes
- Very high mechanical stability
- Clear, scratch-proof, exchangeable non-fog lens
- Wide supporting surface
- Large field of vision
- Maximum wearing comfort
- 99.99% UV protection with innovative UV 380 technology
- Also suitable for people who wear glasses and respiratory masks from EKASTU
Specifications
Specifications
| Certifications/Compliance | DIN EN 166 1 3 4 9 – BT |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Headband Material | Textile Elastic |
| No. per Pack | 1 Ea. |
| Material | Polycarbonate lens |
| Frame Material | PVC |
| UV Protection | Yes |