Learn More
FUT8 Mouse anti-Human, Clone: 2F7H2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66118-1-IG-150UL
This item is not returnable.
View return policy
Description
Catalyzes the addition of fucose in alpha 1-6 linkage to the first GlcNAc residue, next to the peptide chains in N-glycans.Specifications
| FUT8 | |
| Monoclonal | |
| 1.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BYC5 | |
| FUT8 | |
| FUT8 Fusion Protein Ag18769 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2F7H2 | |
| Unconjugated | |
| FUT8 | |
| Alpha (1,6) fucosyltransferase, alpha1 6FucT, Fucosyltransferase 8, FUT8 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2530 | |
| -20°C | |
| Liquid |