missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT8 Mouse anti-Human, Clone: 2F7H2, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | FUT8 |
|---|---|
| Clone | 2F7H2 |
| Concentration | 1.07 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837705
|
Proteintech
66118-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827705
|
Proteintech
66118-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyzes the addition of fucose in alpha 1-6 linkage to the first GlcNAc residue, next to the peptide chains in N-glycans.Specifications
| FUT8 | |
| 1.07 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Alpha (1,6) fucosyltransferase, alpha1 6FucT, Fucosyltransferase 8, FUT8 | |
| FUT8 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2F7H2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BYC5 | |
| 2530 | |
| FUT8 Fusion Protein Ag18769 | |
| Primary | |
| -20°C | |
| FUT8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title