missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha inhibitor 1/2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3522-0.025mg
This item is not returnable.
View return policy
Description
G protein alpha inhibitor 1/2 Polyclonal specifically detects G protein alpha inhibitor 1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Immunofluorescence.
Specifications
| G protein alpha inhibitor 1/2 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 1mg/ml BSA with 0.05% Sodium Azide | |
| GNAI1 | |
| Synthetic peptide corresponding to residues K(346) N N L K D C G L F(355) of human G Protein alpha Inhibitor 1/2. | |
| 0.025 mg | |
| Primary | |
| NB120-3522 detects G protein alpha inhibitor 1 (Gene ID: 2770, UniProt: P63096), and G protein alpha inhibitor 2 (Gene ID: 2771, UniProt: P04899) from human, rat, and mouse tissues and cells. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunofluorescence | |
| GIP, GNAI1, GNAI1/2, GNAI2, GNAI2B, H_LUCA15.1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2770 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction