missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha inhibitor 1/2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 200.00
Specifications
| Antigen | G protein alpha inhibitor 1/2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunofluorescence |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
G protein alpha inhibitor 1/2 Polyclonal specifically detects G protein alpha inhibitor 1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Immunofluorescence.Specifications
| G protein alpha inhibitor 1/2 | |
| Western Blot 1 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunofluorescence | |
| Polyclonal | |
| Rabbit | |
| Human | |
| GIP, GNAI1, GNAI1/2, GNAI2, GNAI2B, H_LUCA15.1 | |
| GNAI1 | |
| IgG | |
| Affinity Purified | |
| NB120-3522 detects G protein alpha inhibitor 1 (Gene ID: 2770, UniProt: P63096), and G protein alpha inhibitor 2 (Gene ID: 2771, UniProt: P04899) from human, rat, and mouse tissues and cells. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS containing 1mg/ml BSA with 0.05% Sodium Azide | |
| 2770 | |
| Synthetic peptide corresponding to residues K(346) N N L K D C G L F(355) of human G Protein alpha Inhibitor 1/2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title