missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04453-100UL
This item is not returnable.
View return policy
Description
GAB1 Polyclonal specifically detects GAB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GAB1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| GRB2-associated binder 1, GRB2-associated binding protein 1, GRB2-associated-binding protein 1, Growth factor receptor bound protein 2-associated protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GAB1 (NP_997006.1). | |
| 100 μL | |
| Apoptosis, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| 2549 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction