missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 483.00
Specifications
| Antigen | GAB1 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18373377
|
Novus Biologicals
NBP3-04453-20UL |
20 μL |
€ 204.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18025686
|
Novus Biologicals
NBP3-04453-100UL |
100 μL |
€ 483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAB1 Polyclonal specifically detects GAB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GAB1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| GRB2-associated binder 1, GRB2-associated binding protein 1, GRB2-associated-binding protein 1, Growth factor receptor bound protein 2-associated protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GAB1 (NP_997006.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| PBS with 50% glycerol, pH7.3. | |
| 2549 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title