missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-9 Antibody (RG9-35) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-09033-0.2mg
This item is not returnable.
View return policy
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Flow Cytometry, Neutralization, Functional Assay
Specifications
| Galectin-9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Neutralization, Functional Assay | |
| RG9-35 | |
| Unconjugated | |
| ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
| This antibody was raised by immunising rats with recombinant mouse galectin-9. | |
| 0.2 mg | |
| Myeloid derived Suppressor Cell, Signal Transduction | |
| 3965 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction