missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-9 Antibody (RG9-35) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 250.00 - € 680.00
Specifications
| Antigen | Galectin-9 |
|---|---|
| Clone | RG9-35 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Neutralization, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682717
|
Novus Biologicals
NBP3-09033-0.2mg |
0.2 mg |
€ 680.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647056
|
Novus Biologicals
NBP3-09033-0.025mg |
0.025 mg |
€ 250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Flow Cytometry, Neutralization, Functional AssaySpecifications
| Galectin-9 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
| This antibody was raised by immunising rats with recombinant mouse galectin-9. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| RG9-35 | |
| Flow Cytometry, Neutralization, Functional Assay | |
| Unconjugated | |
| Rat | |
| Myeloid derived Suppressor Cell, Signal Transduction | |
| PBS | |
| 3965 | |
| IgG2a κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title