missing translation for 'onlineSavingsMsg'
Learn More

Galeterone, MedChemExpress

Product Code. 16461336
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461336

Brand: Medchem Express HY70006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galeterone (TOK-001) is a multifunctional antiandrogen and CYP17 inhibitor (IC50=47 nM) in castration resistant prostate cancer (CRPC).

Galeterone (TOK-001) is a multifunctional antiandrogen and CYP17 inhibitor (IC50=47 nM) in castration resistant prostate cancer (CRPC).

Specifications

Chemical Name or Material Galeterone
CAS 851983-85-2
Color White
Purity Grade Notes Research
Molecular Formula C26H32N2O
Quantity 200 mg
Synonym TOK-001 VN-124-1
Solubility Information DMSO : 25 mg/mL (64.34 mM; Need ultrasonic)
SMILES C[C@@]12C(N3C=NC4=CC=CC=C34)=CC[C@]1([C@@]5(CC=C6[C@@](C)([C@]5(CC2)[H])CC[C@@H](C6)O)[H])[H]
Molecular Weight (g/mol) 388.55
Formula Weight 388.55
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.