missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALT Antibody (SR1733), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22582-100ul
This item is not returnable.
View return policy
Description
GALT Monoclonal antibody specifically detects GALT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| GALT | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1733 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| EC 2.7.7.12, Gal-1-P uridylyltransferase, galactose-1-phosphate uridyl transferase, galactose-1-phosphate uridylyltransferase, UDP-glucose--hexose-1-phosphate uridylyltransferase | |
| A synthesized peptide derived from human GALT (Uniprot #: P07902) | |
| 100 μg | |
| metabolism | |
| 2592 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction