missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALT Antibody (SR1733), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | GALT |
|---|---|
| Clone | SR1733 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641265
|
Novus Biologicals
NBP3-22582-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627474
|
Novus Biologicals
NBP3-22582-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GALT Monoclonal antibody specifically detects GALT in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| GALT | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.7.12, Gal-1-P uridylyltransferase, galactose-1-phosphate uridyl transferase, galactose-1-phosphate uridylyltransferase, UDP-glucose--hexose-1-phosphate uridylyltransferase | |
| A synthesized peptide derived from human GALT (Uniprot #: P07902) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1733 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2592 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title