Learn More
Invitrogen™ GALT Recombinant Rabbit Monoclonal Antibody (23GB5820), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552920
This item is not returnable.
View return policy
Description
Galactose-1-phosphate uridyl transferase (GALT) catalyzes the second step of the Leloir pathway of galactose metabolism, namely the conversion of UDP-glucose + galactose-1-phosphate to glucose-1-phosphate + UDP-galactose. The absence of this enzyme results in classic galactosemia in humans and can be fatal in the newborn period if lactose is not removed from the diet. The pathophysiology of galactosemia has not been clearly defined.
Specifications
| GALT | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Galt | |
| AW553376; Gal-1-P uridylyltransferase; galactose-1-phosphate uridyl transferase; galactose-1-phosphate uridylyltransferase; galactose-1-phosphate uridylyltransferase isoform 1; GALT; IL11RA; interleukin 11 receptor, alpha; UDP-glucose--hexose-1-phosphate uridylyltransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14430, 2592 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5820 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07902, Q03249 | |
| Galt | |
| A synthesized peptide derived from human GALT (50-90AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |