missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALT Recombinant Rabbit Monoclonal Antibody (23GB5820), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | GALT |
|---|---|
| Clone | 23GB5820 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089534
![]() |
Invitrogen™
MA552920 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089835
|
Invitrogen™
MA552921 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GALT Recombinant Monoclonal antibody specifically detects GALT in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GALT | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P07902, Q03249 | |
| 14430, 2592 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5820 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AW553376; Gal-1-P uridylyltransferase; galactose-1-phosphate uridyl transferase; galactose-1-phosphate uridylyltransferase; galactose-1-phosphate uridylyltransferase isoform 1; GALT; IL11RA; interleukin 11 receptor, alpha; UDP-glucose--hexose-1-phosphate uridylyltransferase | |
| A synthesized peptide derived from human GALT (50-90AA). | |
| Primary | |
| -20°C | |
| Galt |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title