missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamma Adaptin Antibody (SR1005), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22376-100ul
This item is not returnable.
View return policy
Description
Gamma Adaptin Monoclonal antibody specifically detects Gamma Adaptin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Gamma Adaptin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1005 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Adapter-related protein complex 1 subunit gamma-1, Adaptor protein complex AP-1 subunit gamma-1, adaptor-related protein complex 1, gamma 1 subunit, ADTGclathrin assembly protein complex 1 gamma large chain, AP-1 complex subunit gamma-1, CLAPG1clathrin-associated/assembly/adaptor protein, large, gamma 1, Clathrin assembly protein complex 1 gamma-1 large chain, gamma adaptin, gamma1-adaptin, golgi adaptor HA1/AP1 adaptin gamma subunit, Golgi adaptor HA1/AP1 adaptin subunit gamma-1, MGC18255 | |
| A synthesized peptide derived from human Gamma Adaptin (Uniprot #: O43747) | |
| 100 μg | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| 164 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction