missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamma Adaptin Antibody (SR1005), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Gamma Adaptin |
|---|---|
| Clone | SR1005 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611775
|
Novus Biologicals
NBP3-22376-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673145
|
Novus Biologicals
NBP3-22376-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gamma Adaptin Monoclonal antibody specifically detects Gamma Adaptin in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpécification
| Gamma Adaptin | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Adapter-related protein complex 1 subunit gamma-1, Adaptor protein complex AP-1 subunit gamma-1, adaptor-related protein complex 1, gamma 1 subunit, ADTGclathrin assembly protein complex 1 gamma large chain, AP-1 complex subunit gamma-1, CLAPG1clathrin-associated/assembly/adaptor protein, large, gamma 1, Clathrin assembly protein complex 1 gamma-1 large chain, gamma adaptin, gamma1-adaptin, golgi adaptor HA1/AP1 adaptin gamma subunit, Golgi adaptor HA1/AP1 adaptin subunit gamma-1, MGC18255 | |
| A synthesized peptide derived from human Gamma Adaptin (Uniprot #: O43747) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1005 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 164 | |
| IgG | |
| Affinity purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit