missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Mouse anti-Hamster, Human, Clone: 9D6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60245-100UL
This item is not returnable.
View return policy
Description
gamma Tubulin Monoclonal specifically detects gamma Tubulin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| gamma Tubulin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Hamster | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 9D6 | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| TUBG1 | |
| Recombinant human TUBG1 (1-451aa) purified from E. coli. | |
| 100 μL | |
| Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers | |
| 7283 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction