missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Mouse anti-Hamster, Human, Clone: 9D6, Novus Biologicals™
Mouse Monoclonal Antibody
€ 304.00 - € 466.00
Specifications
| Antigen | gamma Tubulin |
|---|---|
| Clone | 9D6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221113
|
Novus Biologicals
NBP2-60245-50UL |
50 μL |
€ 304.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18091826
|
Novus Biologicals
NBP2-60245-100UL |
100 μL |
€ 466.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma Tubulin Monoclonal specifically detects gamma Tubulin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| gamma Tubulin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 7283 | |
| Recombinant human TUBG1 (1-451aa) purified from E. coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 9D6 | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Hamster | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| TUBG1 | |
| IgG2b κ | |
| Protein A purified |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title