Learn More
Ansell™ Gammex™ PF DermaPrene™ Powder-Free Neoprene Surgical Gloves
Product Code. p-8005271
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
Unit Size:
100 pairs
25 pairs
Each
Pack of 100
Product Code. 13206544
Brand: Ansell™ 353202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Powder-free neoprene surgical gloves
  • Provide a secure grip on instruments for easy sure handling in both wet and dry conditions
  • Free of latex proteins and accelerators
  • Minimises allergy risk
  • Outstanding comfort and excellent pinhole and tear resistance
  • An extra-length cuff with an adhesive band provides improved grip on the sleeve and prevents roll-down for a secure fit
  • Excellent resistance to fats, alcohol and a broad range of chemical products
  • Strong resistance to viral penetration
TRUSTED_SUSTAINABILITY
Specifications
Size 6