Learn More
GAP-43 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-92714SS
 View more versions of this product
Product Code. 18067096
  • € 234.00 / 25µL
Description
GAP-43 Polyclonal specifically detects GAP-43 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAP-43
Polyclonal
Western Blot 1:100-1:5000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Chicken
43 kDa
0.025 mL
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience
2596
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgY
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Supplied as an aliquot of IgY preparation with 5mM Sodium Azide
GAP43
C-terminal peptide of rodent GAP43, KEDPEADQEHA coupled to KLH.
SDS-PAGE
RUO
Primary
Human, Mouse, Rat, Pig, Bovine, Chicken
Precipitation