missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 2 publications
€ 234.00 - € 500.00
Specifications
| Antigen | GAP-43 |
|---|---|
| Dilution | Western Blot 1:100-1:5000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067096
|
Novus Biologicals
NBP1-92714SS |
0.025 mL |
€ 234.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18011784
|
Novus Biologicals
NBP1-92714 |
0.1 mL |
€ 500.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAP-43 Polyclonal specifically detects GAP-43 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| GAP-43 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience | |
| Supplied as an aliquot of IgY preparation with 5mM Sodium Azide | |
| 2596 | |
| C-terminal peptide of rodent GAP43, KEDPEADQEHA coupled to KLH. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:100-1:5000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000 | |
| Polyclonal | |
| Precipitation | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Chicken | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| GAP43 | |
| IgY | |
| SDS-PAGE | |
| 43 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title