missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBA Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803364S
This item is not returnable.
View return policy
Description
GBA Monoclonal antibody specifically detects GBA in Human samples. It is validated for Western BlotSpecifications
| GBA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04062 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2629 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1D8 | |
| Unconjugated | |
| GBA | |
| acid beta glucosidase; acid beta-glucosidase; Alglucerase; betaGC; beta-GC; Beta-glucocerebrosidase; Cholesterol glucosyltransferase; Cholesteryl-beta-glucosidase; D-glucosyl-N-acylsphingosine glucohydrolase; Gba; GBA1; GBAP; GBAP1; GC; GCase; GCB; GLUC; glucocerebrosidase; glucosidase, beta, acid; glucosylceramidase; glucosylceramidase beta; glucosylceramidase beta pseudogene 1; glucosylceramidase-like protein; imiglucerase; Lysosomal acid GCase; Lysosomal acid glucosylceramidase; lysosomal glucocerebrosidase; SGTase | |
| Human recombinant protein fragment corresponding to amino acids 40-315 of human GBA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction