missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35306-20ul
This item is not returnable.
View return policy
Description
GCN1L1 Polyclonal antibody specifically detects GCN1L1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| GCN1L1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, ELISA | |
| GCN1, GCN1 (general control of amino-acid synthesis 1, yeast)-like 1, GCN1 general control of amino-acid synthesis 1-like 1 (yeast), GCN1L, GCN1-like protein 1, HsGCN1, KIAA0219hsGCN1, peroxisome proliferator activated receptor interacting complex protein, PRIC295, translational activator GCN1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2462-2671 of human GCN1L1 (NP_006827.1).,, Sequence:, LSAVLQQCLLADVSGIDWMVRHGRSLALSVAVNVAPGRLCAGRYSSDVQEMILSSATADRIPIAVSGVRGMGFLMRHHIETGGGQLPAKLSSLFVKCLQNPSSDIRLVAEKMIWWANKDPLPPLDPQAIKPILKALLDNTKDKNTVVRAYSDQAIVNLLKMRQGEEVFQSLSKILDVASLEVLNEVNRRSLKKLASQADSTEQVDDTILT | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10985 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction