missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 724.00
Specifications
| Antigen | GCN1L1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229103
|
Novus Biologicals
NBP3-35306-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229660
|
Novus Biologicals
NBP3-35306-100ul |
100 μL |
€ 724.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCN1L1 Polyclonal antibody specifically detects GCN1L1 in Human samples. It is validated for ELISA,Western BlotSpecifications
| GCN1L1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| GCN1, GCN1 (general control of amino-acid synthesis 1, yeast)-like 1, GCN1 general control of amino-acid synthesis 1-like 1 (yeast), GCN1L, GCN1-like protein 1, HsGCN1, KIAA0219hsGCN1, peroxisome proliferator activated receptor interacting complex protein, PRIC295, translational activator GCN1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2462-2671 of human GCN1L1 (NP_006827.1).,, Sequence:, LSAVLQQCLLADVSGIDWMVRHGRSLALSVAVNVAPGRLCAGRYSSDVQEMILSSATADRIPIAVSGVRGMGFLMRHHIETGGGQLPAKLSSLFVKCLQNPSSDIRLVAEKMIWWANKDPLPPLDPQAIKPILKALLDNTKDKNTVVRAYSDQAIVNLLKMRQGEEVFQSLSKILDVASLEVLNEVNRRSLKKLASQADSTEQVDDTILT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:5000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10985 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title