Learn More
Invitrogen™ GCN2 Recombinant Rabbit Monoclonal Antibody (23GB5295), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553880
 View more versions of this product
Product Code. 30089804
  • € 562.00 / 100µL
Description
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GCN2
Recombinant Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
D4A7V9, Q9P2K8
EIF2AK4
A synthesized peptide derived from human GCN2 (1-59AA).
100 μL
Primary
Human, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5295
Unconjugated
EIF2AK4
2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2
Rabbit
Affinity chromatography
RUO
114859, 440275
-20°C