missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCN2 Recombinant Rabbit Monoclonal Antibody (23GB5295), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | GCN2 |
|---|---|
| Clone | 23GB5295 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
30089804
![]() |
Invitrogen™
MA553880 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088218
|
Invitrogen™
MA553881 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCN2 Recombinant Monoclonal antibody specifically detects GCN2 in Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpécification
| GCN2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| D4A7V9, Q9P2K8 | |
| 114859, 440275 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5295 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2 | |
| A synthesized peptide derived from human GCN2 (1-59AA). | |
| Primary | |
| -20°C | |
| EIF2AK4 |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel