missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GDC 0879
GREENER_CHOICE

Product Code. 18024862
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18024862

Brand: Tocris Bioscience™ 4453/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent B-Raf inhibitor

Potent and selective B-Raf inhibitor (IC50 = 0.13 nM against purified B-Raf V600E). Activity reduces phospho-ERK (pERK) levels (IC50 = 63 nM in the Malme-3M cell line). Inhibits the Raf/MEK/ERK signaling pathway in V600E B-Raf mutant cell lines. Does not activate apoptotic pathways in A375 or Colo205 cell lines. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 905281-76-7
Molecular Formula C19H18N4O2

Specifications

Quantity 10 mg
Formula Weight 334.37
Purity >98%
Molecular Weight (g/mol) 334.379
Chemical Name or Material GDC 0879
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.