missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GDF-11/GDF-8 Antibody (lintuzumAb) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | GDF-11/GDF-8 |
|---|---|
| Clone | lintuzumAb |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044826
|
Novus Biologicals™
NBP3-28789-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045385
|
Novus Biologicals™
NBP3-28789-100ug |
100 μg |
€ 326.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044230
|
Novus Biologicals™
NBP3-28789-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-11/GDF-8 Monoclonal antibody specifically detects GDF-11/GDF-8 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| GDF-11/GDF-8 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Human | |
| O14793 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| lintuzumAb | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| GDF8/ Myostatin | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title