missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin/GSN Antibody (GS-2C4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB110-10067SS
This item is not returnable.
View return policy
Description
Gelsolin/GSN Monoclonal specifically detects Gelsolin/GSN in Human, Porcine, Bovine, Rabbit, Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.
Specifications
| Gelsolin/GSN | |
| Monoclonal | |
| Unconjugated | |
| Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type) | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| The antibody is specific for an epitope located on the 47 kDa peptide derived from a chymotryptic cleavage of gelsolin. The antibody reacts with plasma and cytoplasmic gelsolin | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| GS-2C4 | |
| Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Microarray, Immunoaffinity Purification | |
| GSN | |
| Full length native protein (purified) (Human). | |
| 0.02 mL | |
| Apoptosis, Cancer, Cytoskeleton Markers, Tumor Suppressors | |
| 2934 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction