missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin/GSN Antibody (GS-2C4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
€ 191.00
Specifications
| Antigen | Gelsolin/GSN |
|---|---|
| Clone | GS-2C4 |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Microarray, Immunoaffinity Purification |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Gelsolin/GSN Monoclonal specifically detects Gelsolin/GSN in Human, Porcine, Bovine, Rabbit, Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.Specifications
| Gelsolin/GSN | |
| Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Microarray, Immunoaffinity Purification | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 2934 | |
| Full length native protein (purified) (Human). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| GS-2C4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cytoskeleton Markers, Tumor Suppressors | |
| Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type) | |
| GSN | |
| IgG1 | |
| Unpurified | |
| The antibody is specific for an epitope located on the 47 kDa peptide derived from a chymotryptic cleavage of gelsolin. The antibody reacts with plasma and cytoplasmic gelsolin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title