missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN2 Antibody (SR1523), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22356-50ul
This item is not returnable.
View return policy
Description
GEMIN2 Monoclonal antibody specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| GEMIN2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1523 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1 | |
| A synthesized peptide derived from human GEMIN2 (Uniprot #: O14893) | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8487 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction