missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN2 Antibody (SR1523), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | GEMIN2 |
|---|---|
| Clone | SR1523 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661845
|
Novus Biologicals
NBP3-22356-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632354
|
Novus Biologicals
NBP3-22356-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GEMIN2 Monoclonal antibody specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| GEMIN2 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1 | |
| A synthesized peptide derived from human GEMIN2 (Uniprot #: O14893) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1523 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 8487 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title