missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ DNA Trace Bone Buffer Set
Click to view available options
Quantity:
Sufficient for 25 Preparations
Unit Size:
1 set
Description
NucleoSpin™ DNA Trace is designed for the preparation of highly pure genomic DNA from small amounts of any tissue, cell or forensic sample. The NucleoSpin™ funnel columns (NucleoSpin™ DNA Trace F columns) included in the kit are ideally suited for collecting traces of nucleic acids from large volumes. The optimised shape of the NucleoSpin™ funnel columns allows the combination of large volume capacity and small diameter of the binding membrane. The DNA isolated by NucleoSpin™ DNA Trace is ready to use for common downstream applications like PCR*, real time PCR* or other enzymatic reactions. The silica membrane is optimised for the reversible binding of very small quantities of nucleic acids from dilute samples and DNA amounts in the nanogram range can be processed with high recovery. The membrane can also bind up to 25μg of genomic DNA. The small diameter of the silica membrane allows elution volumes as low as 50μL to 100μL of low-salt buffer or water leading to highly enriched samples. 8 to 12 NucleoSpin™ funnel columns can be processed in parallel depending on the centrifuge rotor used. The risk of cross contamination is eliminated by placing the funnel columns into 50 mL polyethylene tubes which can be closed with screw caps, resulting in self contained systems. For elution, 0.5mL elution tubes are fitted directly to the outlet of the funnel columns. For isolation of genomic DNA from human bones additional buffers are necessary – the customised NucleoSpin™ DNA Trace Bone buffer set is required. Kit components: NucleoSpin™ funnel columns, 50 mL polyethylene tubes with screw cap, 0.5mL elution tubes, buffers, proteinase K.
- Silica membrane technology
- Sample size: 10mg tissue, <105 cells
- Format: funnel columns
- Typical yield > 10ng
- Typical recovery > 70%
- Elution volume: 100μL
- Preparation time: 60 min./prep
- Binding capacity: 20μg
- Typical sensitivity: ≥ 1ng Preparation of highly pure genomic DNA from small amounts of any tissue, cells and forensic samples e.g., dried blood spots.
Specifications
Specifications
| For Use With (Application) | For Isolation of DNA from bones, Only Applicable with NucleoSpin DNA Trace kits, Sufficient for 25 Preparations |
| Quantity | Sufficient for 25 Preparations |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction