missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Wash Buffer PW2 Concentrate
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
Includes two optimised, alternative lysis buffers based on the established CTAB and SDS lysis methods. As plants are very heterogenous and contain many different metabolites such as polyphenols, polysaccharides, or acidic components, NucleoSpin™ Plant II offers two different lysis proceudres for optimal processing of various samples. The silica membrane in the NucleoSpin™ Plant II columns is optimised to improve DNA binding. NucleoSpin™ filters ar included for convenient clarification of lysates.NucleoSpin™ Plant II allows processing of up to 100 mg (wet weight) or 20mg (dry weight) starting material. Depending on the individual sample typical yields are in the range from 1μg to 30μg DNA. The eluted DNA is ready to use for subsequent reactions such as PCR*, restriction analysis and others.Kit components: NucleoSpin™ Plant II columns, filters, collecting tubes 2mL, buffers, RNase A.
Silica membrane technology
- Yield: 1μg to 30μg
- Sample size: ≤100 mg wet weight ≤20mg dry weight
- Elution volume: 100μL
- Binding capacity: 50μg
- Preparation time: 30 min./prep
- NucleoSpin filters included for clarification of lysates
- RNase A included
- Format: mini spin column
- Two alternative lysis buffers included for processing of various samples (PL1 uses CTAB lysis method, PL2 uses SDS lysis method)
Specifications
Specifications
| For Use With (Equipment) | For 250mL Buffer PW2 |
| Quantity | 50 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction