missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rh2, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Ginsenoside Rh2 induces the activation of caspase-8 and caspase-9. Ginsenoside Rh2 induces cancer cell apoptosis in a multi-path manner.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rh2 |
| CAS | 78214-33-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H62O8 |
| Quantity | 10 mg |
| Synonym | 20(S)-Ginsenoside Rh2 20(S)-Rh2 Ginsenoside-Rh2 |
| Solubility Information | DMSO : 50 mg/mL (80.27 mM; Need ultrasonic) |
| SMILES | C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)C)(CC[C@@]2([H])C3(C)C)[C@]5([C@@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC5)C |
| Molecular Weight (g/mol) | 622.87 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction