missing translation for 'onlineSavingsMsg'
Learn More

Gravis Amber Glass Sample Bottles

Product Code. 12506229
Change view
Click to view available options
Color:
Amber
Unit Size:
Pack of 210
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12506229 Amber Pack of 210
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12506229 Supplier Gravis Supplier No. E010251210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Amber pillbox style glass bottles, diameter 30 mm.

  • Amber glass

Specifications

Color Amber
Material Glass
Sterility Non-sterile
Disposable Reusable
Volume (Metric) 25 mL
Graduated No
Style Round
Volume (English) 0.8 oz.
Quantity 210 Pk.
Marking Spot No
Fit for Purpose Certified Quality
Autoclavable Autoclavable
Product Type Sample Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.