Learn More
Hofmann Klaus™ Glass Weighing Boats With Stem
Brand:  Hofmann Klaus™ 24301.06
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10018710
  • € 32.54 / Each
In Stock
to see stock.
Specifications
3 mL
Weighing Boat
1
Glass
Ideal for lossless pouring of the sample
65 mm